Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-2-Triol Solution, 50 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 34114-98-2 |
| Flash Point | 6°C - Closed Cup |
| Linear Formula | C20H30O7 |
| Synonym | 3 alpha,4 beta,15-Trihydroxy-8 alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 382.45 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 μg/mL in acetonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction