Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T- and B-Cell Activation Antigen Rat anti-Mouse, FITC, Clone: GL7, BD

Catalog No. BDB553666 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553666 0.5 mg
BDB562080 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553666 Supplier BD Biosciences Supplier No. 553666
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The GL7 antibody reacts with a 35kDa cell-surface protein found on T and B lymphocytes activated in vitro, on bone marrow Pre-B-II cells, germinal-center B cells, and a subpopulation of the thymocyte fraction expressing high density of CD3e. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6.

Host Species: Rat
Clone: GL7
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen T- and B-Cell Activation Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone GL7
Concentration 0.5mg/mL
Conjugate FITC
Description GL7
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.