Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T- and B-Cell Activation Antigen Rat anti-Mouse, Human, Rat (Reported), BV421, Clone: GL7, BD Horizon™

Catalog No. BDB568851 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568851 0.1 mg
BDB568852 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568851 Supplier BD Biosciences Supplier No. 568851
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

T- and B-Cell Activation Antigen Monoclonal antibody specifically detects T- and B-Cell Activation Antigen in Mouse, Human, Rat (Reported) samples. It is validated for Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen T- and B-Cell Activation Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone GL7
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias GL7; GL-7; GL7 Antigen
Host Species Rat
Immunogen In vitro-activated T-cell-depleted CBA/Ca mouse splenocytes
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Human, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.