Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet Mouse anti-Human, Mouse, BUV395, Clone: O4-46, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568109
This item is not returnable.
View return policy
Description
T-bet Monoclonal antibody specifically detects T-bet in Human, Mouse samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)
Specifications
T-bet | |
Monoclonal | |
Brilliant Ultraviolet 395 | |
T-box expressed in T cells; TBX21; T-box 21; TBLYM | |
Human T-bet Peptide | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
O4-46 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction