Learn More
T-bet/TBX21 Antibody (4B10), Alexa Fluor™ 700, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP143298AF700
Catalog No. NBP143298Z
Only null left
Add to Cart
Description
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).
Specifications
Specifications
T-bet/TBX21
Monoclonal
Alexa Fluor 700
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM
Mouse
Affinity Purified
RUO
30009
Store at 4C in the dark.
Western Blot, Flow Cytometry
4B10
Western Blot, Flow Cytometry, Flow (Intracellular)
TBX21
The immunogen for this antibody was TBet.
0.1 mL
Primary
Human, Mouse, Rhesus Macaque
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only