Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet/TBX21 Antibody (4B10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143298G
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).Specifications
T-bet/TBX21 | |
Monoclonal | |
DyLight 488 | |
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
Mouse | |
Affinity Purified | |
RUO | |
30009 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry | |
4B10 | |
Western Blot, Flow Cytometry, Flow (Intracellular) | |
TBX21 | |
The immunogen for this antibody was TBet. | |
0.1 mL | |
Primary | |
Human, Mouse, Rhesus Macaque | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction