Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet/TBX21 Antibody (4B10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143298F
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human, Mouse, Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular)Specifications
T-bet/TBX21 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Macaque | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Intracellular Staining) | |
4B10 | |
Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Intracellular) | |
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
The immunogen for this antibody was TBet. | |
0.1 mL | |
Immunology | |
30009 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction