Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-bet/TBX21 Antibody (525831), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB53851C
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
T-bet/TBX21 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
525831 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
E. coli-derived recombinant human T-bet/TBX21, Glu326-Asn535, Accession # Q9UL17 | |
0.1 mL | |
Immunology | |
30009 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction