Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-box 19 Antibody (OTI1C9), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274469IR
Description
T-box 19 Monoclonal specifically detects T-box 19 in Human, Mouse samples. It is validated for Western Blot.Specifications
T-box 19 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TBX19 | |
Recombinant protein fragment corresponding to amino acids 1-238 of human T-box 19 (NP_005140) produced in E.coli. | |
0.1 mL | |
9095.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1C9 | |
Western Blot | |
dJ747L4.1, FLJ26302, FLJ34085, T-box 19, T-box factor, pituitary, T-box protein 19, T-box transcription factor TBX19, TBS 19, TBS19, TPITFLJ34543 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction