Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
t-Butyl4-[4-(4,4,5,5-teramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazinecarboxylate, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
| Assay Percent Range | 90% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 388.31 |
| Chemical Name or Material | tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl) phenyl]piperazine-1-carboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction