Learn More
Promega T Cell Activation Bioassay (IL-2), 1 each
Catalog No. PRJ1651 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRJ1651 1 Each
1 options
Catalog No. PRJ1651 Supplier Promega Supplier No. J1651
The T Cell Activation Bioassays are bioluminescent cell-based assays that overcome the limitations of existing assays and can be used for the discovery and development of novel biologics such as bispecific antibodies and CAR-T cell therapies.
  • Measure Stability and Precision of Biologics Designed to Affect T Cell Responses
  • Prequalified according to ICH guidelines
  • Amenable to high−throughput formats
  • No cell culture required
Specifications
Content And Storage BELOW -140°C
Quantity 1 Each