Learn More
Promega T Cell Activation Bioassay (NFAT), Propagation Model, 1 each
Catalog No. PRJ1601 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRJ1601 1 Each
1 options
Catalog No. PRJ1601 Supplier Promega Supplier No. J1601
Add to Cart
Add to Cart
Request a Quote
The T Cell Activation Bioassay, Propagation Model, provides a biologically relevant and specific MOA-based measure of TCR/CD3 Effector Cells without the complex workflow and variability inherent in primary cell-based assays.
  • Allows Propagation and Banking of TCR/CD3 Effector Cells for Use in T Cell Activation Bioassay
Specifications
Content And Storage BELOW -140°C
Quantity 1 Each