Learn More
BD AbSeq™ γδ T-Cell Receptor Hamster, anti-Mouse, Oligo, Clone: GL3, BD AbSeq™
Shop All BD Biosciences ProductsDescription
The GL3 monoclonal antibody specifically binds to a common epitope of the δ chain of the T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with αβ TCR-bearing T cells. In the mouse, cells expressing the γδ TCR are found in the thymus, intestinal epithelium, epidermis, dermis, pulmonsry epithelium, peritoneum, liver, and peripheral lymphoid organs.
Specifications
Specifications
| Antigen | γδ T-Cell Receptor |
| Applications | Single Cell 3' Sequencing |
| Clone | GL3 |
| Conjugate | Oligo |
| Host Species | Hamster |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.