Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
γδ T-Cell Receptor Mouse anti-Rat, BV786, Clone: V65, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744667
Description
γδ T-Cell Receptor Monoclonal antibody specifically detects γδ T-Cell Receptor in Rat samples. It is validated for Flow Cytometry.
Specifications
γδ T-Cell Receptor | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Rat | |
Purified |
Flow Cytometry | |
V65 | |
Brilliant Violet 786 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction