Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

αβ T-Cell Receptor Mouse anti-Rat, FITC, Clone: R73, BD

Catalog No. BDB554913 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554913 0.5 mg
BDB561672 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554913 Supplier BD Biosciences Supplier No. 554913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R73 antibody reacts with the αβ T-cell Receptor (TCR) found on most peripheral T lymphocytes, intestinal intraepithelial lymphocytes, and thymocytes. It does not react with γδ TCR-bearing cells. Cross-linked R73 mAb induces T-cell differentiation and activation. In vivo treatment with mAb R73 can suppress immune function of peripheral αβ TCR-expressing T cells, and reduce the severity of experimental autoimmune, transplant rejection, and graft-versus-host responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen αβ T-Cell Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone R73
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat T blasts and rat erythrocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.