Learn More
γδ T-Cell Receptor Mouse anti-Rat, RB780, Clone: V65, BD OptiBuild™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | γδ T-Cell Receptor |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | V65 |
| Concentration | 0.2 mg/mL |
| Conjugate | RB780 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | TCR γδ; TCR gamma delta |
| Host Species | Mouse |
| Immunogen | Rat CD3+ αβ TCR- T-cell hybridoma III.89.1.4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.