Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T. gondii Profilin-like Protein Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3860
This item is not returnable.
View return policy
Description
Profilin-like Protein Polyclonal specifically detects Profilin-like Protein in T. gondii samples. It is validated for Western Blot.Specifications
Profilin-like Protein | |
Polyclonal | |
Unconjugated | |
AAX47289 | |
E. coli-derived recombinant T. gondii Profilin-like Protein Ser2-Tyr163 Accession # AAX47289 | |
100 μg | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Goat | |
Affinity Purified | |
RUO | |
Detects T. gondii Profilin-like Protein in direct ELISAs and Western blots. | |
T. gondii | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction