Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ T-Lymphocytes Monoclonal Antibody (MsGp7)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528509
Description
Mouse anti Guinea Pig T Lymphocytes, clone MsGp7, is pan reactive with guinea pig T lymphocytes (>95%) in lymph nodes, and does not react with B lymphocytes.
T Lymphocytes are a subtype of white blood cell that is central to cell-mediated immunity. T Lymphocytes mature in the thymus from thymocytes. The majority of human T cells rearrange their alpha and beta chains on the cell receptor and are termed alpha beta T cells (alpha/beta T cells) and are part of the adaptive immune system.
Specifications
T-Lymphocytes | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Guinea Pig T Lymphocytes. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
MsGp7 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Guinea Pig | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction