Learn More
Stoelting™ T Maze
SDP
Stoelting™ T-maze is used largely in preference and spatial learning tasks.
$1968.00 - $2350.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 2
Catalog Number Mfr. No. Animal Price Quantity & Availability  
Catalog Number Mfr. No. Animal Price Quantity & Availability  
10-000-307
SDP
View Documents
Stoelting™
60181
Mouse
Each for $1,968.00
Only null left
Add to Cart
10-000-330
SDP
View Documents
Stoelting™
60281
Rat
Each for $2,350.00
Only null left
Add to Cart
Description
Description
Animals learn to alternate between arms based on their memory of the previously visited arms or choose an arm based on the reward presented. The simple capital T shape design incorporates a single choice point with only two alternatives.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.