Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
t-Plasminogen Activator/tPA Antibody (32B1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | t-Plasminogen Activator/tPA |
---|---|
Clone | 32B1 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
t-Plasminogen Activator/tPA Monoclonal antibody specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Flow Cytometry, ELISASpecifications
t-Plasminogen Activator/tPA | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator | |
A synthesized peptide derived from Human t-Plasminogen Activator/tPA [UniProt P00750] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
32B1 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Cell Biology, Endocrinology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5327 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title