Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
t-Plasminogen Activator/tPA Antibody (OTI3H3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274568UV
Description
t-Plasminogen Activator/tPA Monoclonal specifically detects t-Plasminogen Activator/tPA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
t-Plasminogen Activator/tPA | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLAT | |
Recombinant protein fragment corresponding to amino acids 157-426 of human t-Plasminogen Activator/tPA (NP_000921) produced in E.coli. | |
0.1 mL | |
5327.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3H3 | |
Western Blot, Immunohistochemistry | |
alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, t-PA, TPA, t-plasminogen activator | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction