Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ O-Desmethyl-cis-tramadol Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
Description
O-Desmethyltramadol is an opioid analgesic and an active primary blood metabolite of the synthetic analgesic tramadol. O-Desmethyltramadol is the active component in the designer drug 'Krypton', which is marketed in many countries as a legal high. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as urine drug testing, pain prescription monitoring, forensic analysis, and clinical toxicology.

Specifications
Specifications
Packaging | Ampule |
CAS | 67-56-1 |
Flash Point | 9.7°C (49.5°F) - closed cup |
Concentration | 1.0 mg/mL in methanol (as free base) |
Linear Formula | C15H24ClNO2 |
MDL Number | MFCD08063709 |
UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
Quantity | 1 mL |
Formula Weight | 285.81 |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction