Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T140 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983881
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3881 Supplier Bioss Supplier No. BS15559R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.

Specifications

Antigen T140
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IFT140
Gene Accession No. Q96RY7
Gene Alias AI661311; c305C8.4; c380F5.1; gs114; IFT140; intraflagellar transport 140; intraflagellar transport 140 homolog; Intraflagellar transport protein 140 homolog; KIAA0590; mKIAA0590; MZSDS; SRTD9; Tce5; T-complex expressed gene 5; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2; WDTC2
Gene Symbols IFT140
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IFT140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9742
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.