Learn More
Description
T3 is a thyroid hormone. Promotes differentiation of oligodendroglial precursor cells in neonatal rats and promotes maturation of hiPSC-derived cardiomyocytes. Also specifies cone cell subtype in retinal cell differentiation protocols. T3 is used in protocols to generate cortical spheroids from hPSCs.
For more information about how T3 may be used, see our protocol:
Generation of β cells from hPSCsSpecifications
Specifications
| Chemical Name or Material | O-(4-Hydroxy-3-iodophenyl-3,5-diiodo-L-tyrosine |
| CAS | 6893-02-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Compounds for Stem Cell Differentiation |
| Molecular Formula | C15H12I3NO4 |
| Formula Weight | 650.97 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

