Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ T3 CLK
GREENER_CHOICESDP

Catalog No. 757010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-701-0 10 mg
75-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-701-0 Supplier Tocris Bioscience™ Supplier No. 7570/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective pan-CLK inhibitor; also inhibits DYRK1A/B

T3 CLK is a potent and selective pan CDC-like kinase (CLK) inhibitor (IC50 values are 0.67 nM, 15 nM, and 110 nM for CLK1, CLK2, and CLK3 protein kinases, respectively). T3 CLK also inhibits DYRK1A/B (IC50 values are 230 nM and 260 nM for DYRK1B and DYRK1A, respectively in cellular assays). Exhibits dose-dependent alternative splicing effects in HCT116 colorectal cancer cells. T3 CLK is cell permeable, stable and non-toxic.

Specifications

Chemical Name or Material 4-[1,1-Dimethyl-2-(4-methyl-1-piperazinyl)-2-oxoethyl]-N-[6-(4-pyridinyl)imidazo[1,2-a]pyridin-2-yl]benzamide
CAS 2109805-56-1
Quantity 10 mg
Target Cdc2-like Kinase Inhibitors
Molecular Formula C28H30N6O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.