Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T3 Monoclonal Antibody (3A6)
GREENER_CHOICE

Catalog No. MA121669
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA121669 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA121669 Supplier Invitrogen™ Supplier No. MA121669
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-21669 detects Triiodothyronine hormone from most mammals. This antibody detects T3-BSA and free triiodothyronine in ELISA. The cross-reactivity with thyroxine is <0.1%.

T3 is a hormone made by the thyroid gland. It has three iodine molecules attached to its molecular structure. It is the most powerful thyroid hormone, and affects almost every process in the body, including body temperature, growth, and heart rate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T3
Applications ELISA
Classification Monoclonal
Clone 3A6
Concentration 4.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C15H12I3NO4; Iothyronine; Liothyronin; Tresitope; Triiodothyronine
Host Species Mouse
Immunogen Conjugate of purified triiodothyronine with BSA.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.