Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ T3 (Total) ChLIA Kit

Catalog No. p-7162271 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 x 96 Wells
2 x 96 Wells

Quantitative Chemiluminescence Immunoassay

The T3 (Total) Chemiluminescence Immunoassay kit is intended for the measurement of Total Triiodothyronine (T3) in human serum or plasma.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
Quantity 1 x 96 Wells
Regulatory Status FDA Exempt
Sample Volume 50 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.