Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedical™ T3 Uptake ChLIA Kit
Click to view available options
Quantity:
1 x 96 Wells
2 x 96 Wells
Description
The T3 Uptake Chemiluminescence Immunoassay kit is intended for the measurement of the total amount of binding sites available for the Thyroid Hormones in human serum or plasma.

Specifications
Specifications
Product Type | Kit and Assay |
Sample Type | Plasma, Serum |
Target Species | Human |
Quantity | 1 x 96 Wells |
Regulatory Status | FDA Exempt |
Sample Volume | 25 μL |
Storage Requirements | 2°C to 8°C |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction