Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ T4 gp32 Bulk
Click to view available options
:
100μg
Description
The supplied protein is a purified recombinant form free of endonucleases and nickases. The components of the storage buffer are: Tris-HCl 20mm, pH 8.0, NaCl 100mm, EDTA 1mm, DTT 0.5mm, Glycerol 50%.
- Enhances DNA polymerase proofreading activity and PCR fidelity
- Increases yield of large PCR DNA fragments

Specifications
Specifications
Quantity | 100 μg |
Regulatory Status | RUO |
Common Name | T4 bacteriophage gene 32 protein |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction