Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T4 (L-Thyroxine) Mouse, Clone: T4YCH, Abnova™

Catalog No. 89035512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-512 Supplier Abnova Corporation Supplier No. MAB14114

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against T4 (L-Thyroxine).

  • High specificity
  • High reactivity

Specifications

Antigen T4 (L-Thyroxine)
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone T4YCH
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against T4 (L-Thyroxine).
Dilution ELISA Immunoprecipitation Radioimmunoassay The optimal working dilution should be determined by the end user.
Formulation In 0.01 M potassium phosphate buffer, 2.5 mM KCl, 0.135 M NaCl (1 mg/mL BSA, 0.05% sodium azide).
Host Species Mouse
Immunogen Thyroxine-BSA.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.