Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences T4 Polynucleotide Kinase (PNK)
SCI_ED

Catalog No. S03102
Change view
Click to view available options
Quantity:
2500 Units
500 Units
5000 Units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03102 5000 Units
S03104 2500 Units
S03106 500 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03102 Supplier G-Biosciences Supplier No. 7862009
Add to Cart
Add to Cart

T4 Polynucleotide Kinase (PNK) exhibits 5' polynucleotide kinase and 3' phosphatase activity.

  • T4 Polynucleotide Kinase (PNK) catalyzes 5'-phosphorylation of DNA and oligonucleotides via transferring the terminal phosphate of ATP to a 5' hydroxyl group of a nucleic acid.

Specifications

Enzyme Polynucleotide Kinase
Sensitive to Heat Inactivation Yes
Quantity 5000 Units
Product Type Polynucleotide Kinase
Research Category DNA Repair, Next-generation Sequencing, Genome Editing, Traditional Cloning, Scarless Cloning
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.