Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ T4S20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566779 Supplier Invitrogen™ Supplier No. PA566779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHPESFNLQWF FNDSCAPPTG FNKPTSNDTM ASGWRASSFH FDSEENKHRL IHFSV Highest antigen sequence identity to the following orthologs - mouse 52%, rat 52%.

Polytopic transmembrane protein that inhibits regulated intramembrane proteolysis (RIP) of CREB3L1, inhibiting its activation and the induction of collagen synthesis (PubMed:25310401, PubMed:27499293). In response to ceramide, which alters TM4SF20 membrane topology, stimulates RIP activation of CREB3L1 (PubMed:27499293). Ceramide reverses the direction through which transmembrane helices are translocated into the endoplasmic reticulum membrane during translation of TM4SF20, this mechanism is called 'regulated alternative translocation' (RAT) and regulates the function of the transmembrane protein (PubMed:27499293).
TRUSTED_SUSTAINABILITY

Specifications

Antigen T4S20
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TM4SF20
Gene Accession No. Q53R12
Gene Alias PRO994; SLI5; TCCE518; TM4SF20; transmembrane 4 L six family member 20; transmembrane 4 L6 family member 20; UNQ518/PRO994
Gene Symbols TM4SF20
Host Species Rabbit
Immunogen Recombinant protein fragment of Human TM4SF20 (Product #RP-104287)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.