Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific T500 Screw Caps for Internally Threaded Sample Tubes

Catalog No. 23045265 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-265 Natural
23-045-266 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-265 Supplier Simport™ Scientific Supplier No. T500NOS
Add to Cart
Add to Cart
This item is not returnable. View return policy

T500 Screw Caps for Internally Threaded Sample Tubes available with silicone washer seal.

  • Made of polypropylene
  • With silicone washer seal
  • Autoclavable
  • For internally threaded tubes
  • Tubes sold separately

Specifications

Liner Material Silicone
Pre-sterilized No
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
No. per Pack 1000 Pk.
For Use With (Equipment) T500 Series Sample Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.