Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7 Polyclonal Antibody, HRP, QED Bioscience™
SDP

Catalog No. 502277851
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7851 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7851 Supplier Leinco Technologies Inc Supplier No. 18862P1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

T7 Polyclonal antibody specifically detects T7 in Tag samples. It is validated for ELISA, Immunocytochemistry, Western Blot

Specifications

Antigen T7
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.2% BSA and 0.05% Pro-Clean 400; pH 7.2
Gene Alias MASMTGGQQMG; T7 cleavage
Host Species Goat
Immunogen T7 (MASMTGGQQMG) conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.