Learn More
T7-tag Rabbit anti-Recombinant Protein, Unconjugated, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The T7-tag antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of T7-tag. This antibody recognizes recombinant protein antigen. The T7-tag antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | T7-tag |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 200 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.