Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T7-tag Rabbit, Polyclonal, GenScript™

Catalog No. 50272854
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-854 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-272-854 Supplier Genscript Corporation Supplier No. A0062240

Rabbit Polyclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL. Well-characterized antibodies with useful epitope tags have been widely used in the study of protein expression in various systems.The T7 tag (MASMTGGQQMG) is a peptide with 11 amino acids encoded in the leader sequence of T7 bacteriophage gene10. This gene encodes a T7 major capsid protein.The T7 tag serves as a tag in many expression vectors including the pET system that is based on the efficient T7 RNA polymerase expression system.GenScript Rabbit Anti-T7-tag Polyclonal Antibody is developed in rabbit using a synthetic T7-tag epitope peptide conjugated to KLH.

T7-tag Polyclonal antibody specifically detects T7-tag in Tag samples. It is validated for Western Blot

Specifications

Antigen T7-tag
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias MASMTGGQQMG; T7 cleavage
Host Species Rabbit
Immunogen T7-tag epitope peptide conjugated to KLH
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.