Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAAR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599933
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599933 Supplier Invitrogen™ Supplier No. PA599933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAAR2.

Orphan receptor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAAR2
Gene Accession No. Q5QD17, Q5QD25, Q9P1P5
Gene Alias G protein-coupled receptor 58; Gm224; Gm225; GPR58; G-protein coupled receptor 58; mTaar2; PhBL5; TAAR2; taR-2; Tarr2; trace amine associated receptor 2; trace amine receptor 2; trace amine-associated receptor 2
Gene Symbols TAAR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAAR2(Accession Q9P1P5), corresponding to amino acid residues S11-N61.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209512, 294121, 9287
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.