Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAAR3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132858
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132858 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132858 Supplier Thermo Scientific Supplier No. PA132858

Rabbit Polyclonal Antibody

PA1-32858 detects GPCR GPR58 from human samples.

Olfactory receptor activated by several primary trace amines, including isoamylamine. Activated by isoamylamine and cyclohexylamine, but not to the corresponding alcohols, isoamylalcohol and cyclohexanol. This receptor is probably mediated by the G(s)-class of G-proteins which activate adenylate cyclase. [GeneCards]
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAAR3P
Gene Accession No. Q9P1P4
Gene Alias GPR57; GPR58; mTaar3; TAAR2; Taar3; TaR-3; trace amine associated receptor 3; trace amine associated receptor 3 (gene/pseudogene); trace amine receptor 3; trace amine-associated receptor 3
Gene Symbols TAAR3P
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human GPCR GPR58
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.