Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAAR5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA521919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521919 Supplier Thermo Scientific Supplier No. PA521919

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2, mouse muscle. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The photoreceptor-specific nuclear receptor (PNR) is a recently identified nuclear transcription factor. PNR expression pattern appears to be restricted in the retina, specific to photoreceptor cells. PNR has been shown to share structural similarity with TLX, an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR5
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Taar5
Gene Accession No. O14804, Q5QD14
Gene Alias Gm227; hTaar5; MGC138414; MGC138416; mTaar5; PNR; Putative neurotransmitter receptor; RP11-295F4.5; TAAR5; TaR-5; trace amine associated receptor 5; trace amine receptor 5; trace amine-associated receptor 5; Trimethylamine receptor
Gene Symbols Taar5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 273 and 337 of TAAR5 (Uniprot ID#O14804)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215854, 9038
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.