Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAAR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533069
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533069 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533069 Supplier Invitrogen™ Supplier No. PA533069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Galago (83%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed in kidney and amygdala. Not expressed in other tissues or brain regions tested.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR8
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAAR8
Gene Accession No. Q969N4
Gene Alias G protein-coupled receptor 102; Gm1149; Gm230; GPR102; G-protein coupled receptor 102; mTaar8a; Q923; Ta10; Ta11; TA5; Ta7; TAAR8; Taar8a; Taar8b; Taar8c; Tar11; TaR-11; TAR5; TaR-5; TaR-8; TaR-8a; trace amine associated receptor 8; trace amine receptor 11; Trace amine receptor 5; trace amine receptor 8; trace amine receptor 8a; trace amine-associated receptor 8; Trace amine-associated receptor 8a; trace-amine-associated receptor 8a; Trar11; Trar5
Gene Symbols TAAR8
Host Species Rabbit
Immunogen Synthetic 12 amino acid peptide from C-Terminus of human TAAR8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83551
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.