Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAAR9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA120783
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120783 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120783 Supplier Invitrogen™ Supplier No. PA120783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-20783 detects TA3 in Human samples.

TAAR9 is a member of a large family of rhodopsin G protein-coupled receptors (GPCRs, or GPRs). GPCRs contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAAR9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TAAR9
Gene Accession No. Q96RI9
Gene Alias Gm231; mTaar9; SNORF1 receptor; TA3; Taar9; TAR3; TaR-3; TAR9; taR-9; trace amine associated receptor 9 (gene/pseudogene); trace amine receptor 3; trace amine receptor 9; trace amine-associated receptor 9; trace-amine-associated receptor 9; Trar3
Gene Symbols TAAR9
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human TAAR9.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.