Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAB-2 Mouse anti-Human, Clone: 1D5.2, MilliporeSigma™

Catalog No. MABS189425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1894-25 25 μg
MABS1894-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS1894-25 Supplier MilliporeSigma Supplier No. MABS189425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TAB-2, clone 1D5.2 is a mouse monoclonal antibody that detects TAK1-binding protein 2 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAB-2
Applications Western Blot
Classification Monoclonal
Clone 1D5.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TAB2,KIAA0733,MAP3K7IP2
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 133 amino acids from the C-terminal region of human TAK1-binding protein 2 (TAB-2)
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001278963
Test Specificity Clone 1D5.2 specifically detects human TGF-beta-activated kinase 1 and MAP3K7-binding protein 2. It targets an epitope within 133 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.