Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAB182 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558172 Supplier Invitrogen™ Supplier No. PA558172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVLASPDRLW GSRLTFNHDG SSRYGPRTYG TTTAPRDEDG STLFRGWSQE GPVKSPAECR EEHSKTPEER SLPSDLAFNG DLAKAASSEL PAD Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 66%.

TAB182 (tankyrase 1-binding protein of 182 kDa) was identified in a two-hybrid screen in search of tankyrase 1-interacting proteins. Tankyrase 1 is a poly (ADP-ribose) polymerase (PARP) that ribosylates the negative regulator of telomere length, TRF1 (telomere-repeat binding factor). Ribosylation of TRF1 has been shown to result in the inhibition of TRF1 binding and an induction of telomere elongation. TAB182 and TRF1 bind multiple discrete and overlapping sites of the tankyrase 1 ankyrin domain. This finding suggests that TAB182 may act as a scaffold to mediate higher order protein complex formation at telomeres. TAB182 has also been suggested to serve a role in the cytoplasm as it has been found to localize to the cytoplasmic cortical actin network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAB182
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNKS1BP1
Gene Accession No. Q9C0C2
Gene Alias 182 kDa tankyrase-1-binding protein; KIAA1741; TAB182; tankyrase 1 binding protein 1; tankyrase 1 binding protein 1, 182kDa; tankyrase 1-binding protein of 182 kDa; testis secretory sperm-binding protein Li 206a; TNKS1BP1
Gene Symbols TNKS1BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAB182. Recombinant protein control fragment (Product #RP-97361).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.