Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557100
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA557100 100 μL
1 options

Catalog No. PIPA557100

Supplier: Invitrogen™ PA557100

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPKPPFSVNP VYITYTQPTG PSCTPSPSPR VIPNPTTVFK ITVGRATTEN LLNLVDQEER SAAPEPIQPI SVIPGSGGEK GSHKYQRSSS SGSDDYA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TAB3 functions in the NF-kappaB signal transduction pathway. It and the similar and functionally redundant protein TAB2, form a ternary complex with the protein kinase TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. Recent experiments have shown that TAB2 and the related protein TAB3 constitutitvely interact with the autophagy mediator Beclin-1; upon induction of autophagy, these proteins dissociate from Beclin-1 and bind TAK1. Overexpression of TAB2 and TAB3 inhibit autophagy, while their depletion triggers it, suggesting that TAB2 and TAB3 act as a control point for autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAB3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tab3
Gene Accession No. Q8N5C8
Gene Alias 4921526G09Rik; Kiaa4135; MAP3K7IP3; mitogen activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7-interacting protein 3; mKIAA4135; NAP1; NF-kappa-B-activating protein 1; NFkB activating protein 1; Tab3; TAB-3; TAK1-binding protein 3; TGF-beta activated kinase 1 (MAP3K7) binding protein 3; TGF-beta activated kinase 1 and MAP3K7 binding protein 3; TGF-beta activated kinase 1/MAP3K7 binding protein 3; TGF-beta-activated kinase 1 and MAP3K7-binding protein 3; TGF-beta-activated kinase 1-binding protein 3
Gene Symbols Tab3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAB3. Recombinant protein control fragment (Product #RP-95454).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257397
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.