Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tabimorelin hemifumarate
GREENER_CHOICESDP

Catalog No. 230810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-081-0 Supplier Tocris Bioscience™ Supplier No. 2308/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent, orally active ghrelin receptor agonist

Potent, orally active ghrelin receptor (GHS-R1a) agonist (Ki = 50 nM at human recombinant GHS-R1a). Stimulates GH release from rat pituitary cells with an EC50 value of 2.7 nM. Induces hyperphagia and adiposity in lean rats, but not in leptin signaling-deficient ZDF rats.

Specifications

CAS 242143-80-2
Quantity 10 mg
Product Type Tabimorelin hemifumarate
Purity >98%
Molecular Weight (g/mol) 1173.466
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.