Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TABS, 99%

Catalog No. AC428080250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428080250 25 g Glass bottle
AC428081000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428080250 Supplier Thermo Scientific Chemicals Supplier No. 428080250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 54960-65-5257.301 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54960-65-5
Molecular Weight (g/mol) 257.301
Synonym N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Packaging Glass bottle
Percent Purity ≥98.5% (on anhydrous basis)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.