Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TAC1 C-terminal Peptide Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595824
Description
Immunogen sequence: MKILVALAVF FLVSTQLFAE EIGANDDLNY WSDWYDSDQI KEELPEPFEH LLQRIARRPK PQQFFGLMGK RDADSSIEKQ VALLKALYGH GQISHKRHKT DSFVGLMGKR ALNSVAYERS AMQNYERRR; Positive Samples: SW620.
Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocorticoid receptor (GR) and that complex's subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain glucocorticoid receptor.
Specifications
TAC1 C-terminal Peptide | |
Polyclonal | |
Unconjugated | |
TAC1 | |
C-terminal-flanking peptide; Hs.2563; ne; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK2; NKA; NKNA; NPK; PPT; preprotachykinin; protachykinin; protachykinin-1; Substance K; substance P; TAC1; TAC2; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6863 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20366 | |
TAC1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human TAC1 (NP_0031731). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction