Learn More
Invitrogen™ TAC1 C-terminal Peptide Polyclonal Antibody

Description
This antibody is predicted to react with mouse, rat and rabbit based on sequence homology.
Specifications
Specifications
| Antigen | TAC1 C-terminal Peptide |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TAC1 |
| Gene Accession No. | P20366 |
| Gene Alias | C-terminal-flanking peptide; Hs.2563; ne; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK2; NKA; NKNA; NPK; PPT; preprotachykinin; protachykinin; protachykinin-1; Substance K; substance P; TAC1; TAC2; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma) |
| Gene Symbols | TAC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.