Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110792 Supplier Invitrogen™ Supplier No. PA5110792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WSDWYDSDQI KEELPEPFEH LLQRIARRPK PQQFFGLMGK RDADSSIEKQ VALLKALYGH GQISHKRHKT DSFVGLMGKR ALNSVAYERS AMQNYERRR Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 96%.

This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TAC1
Gene Accession No. P20366
Gene Alias 4930528L02Rik; C-terminal-flanking peptide; Hs.2563; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK1; NK-1; NK2; NKA; NKNA; NPK; PPT; Ppt5fl; PPTA; PPT-A; PPTA3; preprotachykinin; preprotachykinin A; protachykinin; protachykinin-1; RATPPTA3; SP; substance K; Substance P; TAC; Tac1; Tac2; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Symbols TAC1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TAC1. Recombinant protein control fragment (Product #RP-90655).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.