Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 138391AP150UL
Description
This antibody is specific to TAC1.
This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| TAC1 | |
| Polyclonal | |
| Unconjugated | |
| TAC1 | |
| Hs.2563, Neuromedin L, NK2, NKA, NKNA, PPT, Protachykinin 1, Substance K Neuropeptide K, Substance P, SubstanceP, TAC1, TAC2, tachykinin, precursor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21333, 6863 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20366, P41539 | |
| TAC1 | |
| TAC1 Fusion Protein Ag4790 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction